Learn More
New England Biolabs, Inc. OneTaq DNA Polymerase – 5000 units
SDP
Catalog No. 504388
Encompass
The 3' to 5' exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. The OneTaq Reaction Buffers and High GC Enhancer have been formulated for robust yields with minimal optimization, regardless of a template's GC content. OneTaq DNA Polymerase is supplied with two 5X buffers: (Standard and GC), as well as a High GC Enhancer solution. For most routine and/or AT-rich amplicons (Lambda, etc.) or complex amplicons with up to ~65% GC content, OneTaq Standard Reaction Buffer provides robust amplification. For GC-rich amplicons, the OneTaq GC Reaction Buffer can improve both performance and yield. For particularly high GC or difficult amplicons, the OneTaq High GC Enhancer can be added at a final concentration of 10-20% to reactions containing OneTaq GC Reaction Buffer.
Catalog No. 50-438-8 Supplier New England Biolabs, Inc. Supplier No. M0480X
The 3' to 5' exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. The OneTaq Reaction Buffers and High GC Enhancer have been formulated for robust yields with minimal optimization, regardless of a template's GC content. OneTaq DNA Polymerase is supplied with two 5X buffers: (Standard and GC), as well as a High GC Enhancer solution. For most routine and/or AT-rich amplicons (Lambda, etc.) or complex amplicons with up to ~65% GC content, OneTaq Standard Reaction Buffer provides robust amplification. For GC-rich amplicons, the OneTaq GC Reaction Buffer can improve both performance and yield. For particularly high GC or difficult amplicons, the OneTaq High GC Enhancer can be added at a final concentration of 10-20% to reactions containing OneTaq GC Reaction Buffer.