Learn More
New England Biolabs, Inc. OneTaq Hot Start Quick-Load 2X Master Mix with GC Buffer – 500 reactions (50 µl vol)
SDP
Catalog No. 504399
Encompass
OneTaq Hot Start Quick-Load 2X Master Mix with GC Buffer is an optimized, ready-to-use blend of Taq and Deep Vent DNA Polymerases combined with an aptamer-based inhibitor. This enzyme blend is ideally suited to PCR applications from GC-rich templates, including pure DNA solutions, bacterial colonies, and cDNA products. The 3' to 5' exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. The hot start nature of the enzyme offers convenience with decreased interference from primer dimers and secondary products. The convenient master mix formulation contains dNTPs, MgSO4, buffer components and stabilizers as well as two commonly used tracking dyes for DNA gels. On a 1% agarose gel in 1X TBE, Xylene Cyanol FF migrates at ~4 kb and Tartrazine migrates at ~10 bp. Both dyes are present in concentrations that do not mask co-migrating DNA bands.
Catalog No. 50-439-9 Supplier New England Biolabs, Inc. Supplier No. M0489L
OneTaq Hot Start Quick-Load 2X Master Mix with GC Buffer is an optimized, ready-to-use blend of Taq and Deep Vent DNA Polymerases combined with an aptamer-based inhibitor. This enzyme blend is ideally suited to PCR applications from GC-rich templates, including pure DNA solutions, bacterial colonies, and cDNA products. The 3' to 5' exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. The hot start nature of the enzyme offers convenience with decreased interference from primer dimers and secondary products. The convenient master mix formulation contains dNTPs, MgSO4, buffer components and stabilizers as well as two commonly used tracking dyes for DNA gels. On a 1% agarose gel in 1X TBE, Xylene Cyanol FF migrates at ~4 kb and Tartrazine migrates at ~10 bp. Both dyes are present in concentrations that do not mask co-migrating DNA bands.