Learn More
New England Biolabs, Inc. OneTaq Quick-Load DNA Polymerase – 2500 units
SDP
Catalog No. 501527860
Encompass
OneTaq Quick-Load DNA Polymerase is an optimized, ready-to-use blend of Taq and Deep Vent DNA Polymerases ideally suited to routine PCR applications from a variety of templates, including pure DNA solutions, bacterial colonies and cDNA products. The 3' to 5' exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. The convenient quick-load buffer contains two commonly used tracking dyes for direct loading of PCR products on DNA gels. On a 1% agarose gel in 1X TBE, Xylene Cyanol FF migrates at ~4 kb and Tartrazine migrates at ~10 bp. Both dyes are present in concentrations that do not mask comigrating DNA bands.
Catalog No. 50-152-7860 Supplier New England Biolabs, Inc. Supplier No. M0509X
OneTaq Quick-Load DNA Polymerase is an optimized, ready-to-use blend of Taq and Deep Vent DNA Polymerases ideally suited to routine PCR applications from a variety of templates, including pure DNA solutions, bacterial colonies and cDNA products. The 3' to 5' exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. The convenient quick-load buffer contains two commonly used tracking dyes for direct loading of PCR products on DNA gels. On a 1% agarose gel in 1X TBE, Xylene Cyanol FF migrates at ~4 kb and Tartrazine migrates at ~10 bp. Both dyes are present in concentrations that do not mask comigrating DNA bands.