Learn More
Thermo Scientific ONGUARD CARTRIDEGE 1CC 48/PK
Supplier:  Thermo Scientific 057089
Encompass
Remove chloride, bromide, and iodide from sample matrices to improve performance of ion chromatography (IC) and facilitate trace analyses. Performed prior to IC analysis, the Thermo Scientific™ Dionex™ OnGuard™ II Ag Cartridge is an off-line sample preparation technique that eliminates these matrix interferences using a silver-form, high-capacity, suffocated, cation-exchange resin. Pretreat a single sample or many samples in series. Use them singly or in series, or pretreat up to 12 samples in parallel using the optional Thermo Scientific™ Dionex™ OnGuard™ Sample Prep Station.
Catalog No. NC0340137