Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dunlop™ Onguard™ Chest Waders with Cleated Outsole

Catalog No. 19073201 Shop All Onguard Industries LLC Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
11
12
Toe Material:
Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Toe Material
19-073201 12 Steel
19-024406 11 Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-073201 Supplier Dunlop™ Supplier No. 860671200
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Superior waterproof protection that resists oil, gas, grease, and saltwater in every condition.

These durable chest waders are made of heavy duty PVC and boast cleated outsoles for excellent flexibility and traction clogging prevention, even in wet or muddy terrain.

  • Chest high, with PVC upper and non-absorbent polyester backing
  • Injection molded, seamless boot construction
  • Steel midsole and adjustable polyester and nylon strap
  • Excellent cold weather flexibility
  • Can be folded or rolled without cracking
  • Top drawstring closure for snug fit
  • Non-corrosive, reinforced suspender buttons
  • Built-in belt loops with adjustable belt
  • Replaceable cushion insoles
  • Made in the USA
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ASTM F2413-11 M I/75 C/75 SD
Closure Type Drawstring
Color Black
For Use With (Application) Utilities, Municipalities, Aquaculture, and Commercial Fishing
Material PVC, Polyester
Seam Type Seamless
Size 12
Sole Features Cleated Outsole, Steel Midsole, Replaceable Cushion Insole
Strap Material Polyester/Nylon
Toe Material Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.