Learn More
Dunlop™ Onguard™ Goliath™ PVC Knee Boots
Catalog No. 19099168F Shop All Onguard Industries LLC Products
Click to view available options
Size:
10
11
12
14
Toe Style:
Plain
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Toe Style
19-099-168F 10 Plain
19-099-168G 11 Plain
19-099-168H 12 Plain
19-099-168K 14 Plain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-099-168F Supplier Dunlop™ Supplier No. 89680 10
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ideal for general industrial and safety applications
  • Constructed of a special urethane/PVC compound with Elvaloy™ and polymeric plasticizers for enhanced strength and high abrasion resistance
  • Superior protection from animal fats (especially poultry), acids, oils, greases, and other chemicals
  • Net-lined for easy on and off, with a removable insole
  • Built-in steel shank for firm arch support
  • Black with a gray Power-Lug outsole
Order Info
Available with or without steel toe in men's sizes only
Compliance
Steel toe meets or exceeds ASTM™ F2413-11 specifications.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ASTM F2413-05
Color Black
Design Pull-on
For Use With (Application) Used in food processing, chemical plants, lawn care, canneries, poultry processing, fish processing, breweries and mining
Height (English) 16 in.
Material Urethane, PVC
Shank Material Steel
Size 10
Sole Features Gray power-lug outsole and removable insole
Toe Style Plain
Resistant To Animal fats, acids, oils, greases and chemicals
Waterproof Yes