Learn More
Thermo Scientific Dionex OnGuard II Ba Cartridges | | 1 EA
Supplier:  Thermo Scientific 057093
Encompass
Remove high concentrations of sulfate from sample matrices, Improve ion chromatography (IC) performance and facilitate low-level ion analysis, Uses a styrene-based, sulfonic acid resin in the barium form
Catalog No. NC1145561