Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dunlop™ Onguard™ DuraPro™ Chemical Resistant Plain Toe Boots with Steel Shank

Catalog No. 18500506 Shop All Onguard Industries LLC Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
9
13
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-500-506 9
18-500-510 13
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-500-506 Supplier Dunlop™ Supplier No. 890859
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

High performance compund provides ideal chemical resistance in the upper and superior abrasion and slip resistance in the outsole

  • Sloped comfort top rim prevents chafing and reduces discomfort when walking, bending and kneeling
  • 360° pull tab for easy donning
  • Steel shank protects and supports arch
  • Larger top circumference for quick and easy pull-on
  • 3/4 in. height inner liner allows  increased flexibility, subtle feel at the calf, and soft lined feel of leg and foot
  • Honeycomb design molded inside the heel to reduce impact when walking
  • Ergonomically designed foot-bed to provide better underfoot support
  • Protective bumpers at rear of boot to prevent premature wear/puncture
  • Extended circumference kick-off ledge
  • Composition: Polyblend Upper; Sureflex Outsole
  • Sole Design: Next Generation Safety-Loc
  • Available in sizes ranging from 3 to 16
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Material Polyblend, Sureflex
Shank Material Steel
Size 9
Sole Features Next Generation Safety-loc, slip-resistant
Toe Style Plain
Resistant To Chemicals
Product Type Chemical resistant Footwear
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.