Learn More
Dunlop™ Onguard™ DuraPro™ Chemical Resistant Steel Toe Boots with Steel Shank
Catalog No. 18500524 Shop All Onguard Industries LLC Products
Click to view available options
Size:
7
8
9
10
11
12
13
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-500-524 13
18-500-518 7
18-500-519 8
18-500-520 9
18-500-521 10
18-500-522 11
18-500-523 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 18-500-524 Supplier Dunlop™ Supplier No. 8908613
This item is not returnable. View return policy
High performance steel toe boot provides ideal chemical resistance in the upper and superior abrasion and slip resistance in the outsole
  • Sloped comfort top rim prevents chafing and reduces discomfort when walking, bending and kneeling
  • 360-degree pull tab for easy donning
  • Larger top circumference for quick and easy pull-on
  • 3/4 in. height inner liner allows  increased flexibility, subtle feel at the calf, and soft lined feel of leg and foot
  • Honeycomb design molded inside the heel to reduce impact when walking
  • Polyblend Upper; Sureflex Outsole
  • Ergonomically designed foot-bed to provide better underfoot support
  • Protective bumpers at rear of boot to prevent premature wear/puncture
  • Extended circumference kick-off ledge
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Material Polyblend, Sureflex
Shank Material Steel shank
Size 13
Sole Features Next Generation Safety-loc
Toe Style Steel toe
Resistant To Chemicals