Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dunlop™ Onguard™ Men's PVC Overshoes with Four-Way Cleated Outsole

Catalog No. 19065520D Shop All Onguard Industries LLC Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-065-520D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-065-520D Supplier Dunlop™ Supplier No. 88020XL00
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Injection-molded PVC shoes for seamless protection

  • Excellent cold weather flexibility
  • Four-way, self-cleaning, slip resistant outsole
  • Kick off lug for easy on and off
  • 10 in. height fits over most work shoes and boots
  • Yellow overshoes are made of Flex-O-Thane PVC

TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
For Use With (Application) Used in construction, farming, utilities and food processing
Height (English) 10 in.
Material Flexothane, PVC
Seam Type Seamless
Size X-Large
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.