Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC OnGuard Rain Bib Overalls, 5X-Large, PVC, Chemical Protection, 1 ea

Catalog No. 5000794842
Encompass

These rain bib overalls are engineered for chemical protection, featuring a robust 0.35 mm thick green PVC material in a 5X-Large size. They are designed with heat-sealed seams for enhanced durability in demanding environments.

  • Garment type: chemical
  • Size: 5X-Large
  • Color: green
  • Material: PVC
  • Inseam length: 33 in
  • Material thickness: 0.35 mm
  • Material weight: 32.0 oz
  • Ankle style: snap
  • Seam style: heat sealed
  • Plain front fly
  • Waist size: 72 in
  • Certification: non-hazardous protection
  • Series: Sanitex
  • Closure type: button
  • Number of pockets: 1

Catalog No. 50-007-94842 Supplier MSC Supplier No. 44664951
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These rain bib overalls are engineered for chemical protection, featuring a robust 0.35 mm thick green PVC material in a 5X-Large size. They are designed with heat-sealed seams for enhanced durability in demanding environments.

  • Garment type: chemical
  • Size: 5X-Large
  • Color: green
  • Material: PVC
  • Inseam length: 33 in
  • Material thickness: 0.35 mm
  • Material weight: 32.0 oz
  • Ankle style: snap
  • Seam style: heat sealed
  • Plain front fly
  • Waist size: 72 in
  • Certification: non-hazardous protection
  • Series: Sanitex
  • Closure type: button
  • Number of pockets: 1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.