Learn More
MSC OnGuard Rain Bib Overalls, Large, Nylon and PVC, 1 ea
Catalog No. 5000687920
Encompass
OnGuard Rain Bib Overalls provide protection for chemical applications, constructed from durable Nylon and PVC material.
  • Bib overalls garment style
  • Chemical garment type
  • Open cuff style
  • Open ankle style
  • Plain front
  • Open wrist style
  • Level C certification
  • Chemtex series
  • Zipper closure
  • One pocket
Catalog No. 50-006-87920 Supplier MSC Supplier No. 09215401
OnGuard Rain Bib Overalls provide protection for chemical applications, constructed from durable Nylon and PVC material.
  • Bib overalls garment style
  • Chemical garment type
  • Open cuff style
  • Open ankle style
  • Plain front
  • Open wrist style
  • Level C certification
  • Chemtex series
  • Zipper closure
  • One pocket