Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC OnGuard Rain Bib Overalls, Large, Yellow, PVC, 1 ea

Catalog No. 5000788063
Encompass

These OnGuard Sitex series Rain Bib Overalls are designed for non-hazardous protection. They feature a durable PVC construction for reliable outdoor use.

  • Large size
  • Yellow color
  • PVC material
  • Inseam length: 30 in
  • Snap ankle style
  • Heat sealed seam style
  • Take up snaps at ankles; plain front fly
  • Waist size: 50 in
  • Material weight: 26.4 oz
  • Material thickness: 0.35 mm
  • Snap closure type
  • Plain front fly type
  • Two pockets
  • Country of origin: China

Catalog No. 50-007-88063 Supplier MSC Supplier No. 99496630
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These OnGuard Sitex series Rain Bib Overalls are designed for non-hazardous protection. They feature a durable PVC construction for reliable outdoor use.

  • Large size
  • Yellow color
  • PVC material
  • Inseam length: 30 in
  • Snap ankle style
  • Heat sealed seam style
  • Take up snaps at ankles; plain front fly
  • Waist size: 50 in
  • Material weight: 26.4 oz
  • Material thickness: 0.35 mm
  • Snap closure type
  • Plain front fly type
  • Two pockets
  • Country of origin: China

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.