Learn More
MSC OnGuard Rain and Chemical-Resistant Bib Overalls, 3X-Large, 1 ea
Catalog No. 5000676996
Encompass
These non-disposable rain and chemical-resistant bib overalls are designed for protection. They feature a 3X-Large size, green color, and are constructed from nylon, PVC, and spunbond materials.
  • Garment style: bib overalls
  • Garment type: chemical
  • Ankle style: open
  • Features: plain front
  • Wrist style: open
  • Certification type: Level C
  • Closure type: zipper
  • Number of pockets: 1
Catalog No. 50-006-76996 Supplier MSC Supplier No. 09215435
These non-disposable rain and chemical-resistant bib overalls are designed for protection. They feature a 3X-Large size, green color, and are constructed from nylon, PVC, and spunbond materials.
  • Garment style: bib overalls
  • Garment type: chemical
  • Ankle style: open
  • Features: plain front
  • Wrist style: open
  • Certification type: Level C
  • Closure type: zipper
  • Number of pockets: 1