Learn More
MSC OnGuard Rain Jacket, X-Large, Green, Flame-Resistant, Chemical-Resistant, 1 ea
Catalog No. 5000770302
Encompass
This rain jacket is constructed from a blend of nylon, polyester, and PVC, designed for protective wear in challenging environments. It offers flame and chemical resistance, making it suitable for various industrial applications.
  • X-Large size
  • Green color
  • Material: polyester, nylon, PVC
  • Flame-resistant
  • Chemical-resistant
  • Chest size: 58 in
  • Zipper closure
  • ASTM D6413 certified
  • Chemtex series
  • One pocket
Catalog No. 50-007-70302 Supplier MSC Supplier No. 09215344
This rain jacket is constructed from a blend of nylon, polyester, and PVC, designed for protective wear in challenging environments. It offers flame and chemical resistance, making it suitable for various industrial applications.
  • X-Large size
  • Green color
  • Material: polyester, nylon, PVC
  • Flame-resistant
  • Chemical-resistant
  • Chest size: 58 in
  • Zipper closure
  • ASTM D6413 certified
  • Chemtex series
  • One pocket