Learn More
MSC OnGuard Rain Suit, 3X-Large, Non-Hazardous, Polyester and PVC, 1 ea
Catalog No. 5000770254
Encompass
This rain suit with pants is designed to provide non-hazardous protection. It features a yellow color and is constructed from durable polyester and PVC materials, suitable for various outdoor work environments.
  • Suit with pants garment style
  • Size: 3X-Large
  • Yellow color
  • Polyester and PVC material
  • Non-hazardous protection standards
  • Detachable hood
  • Bib pant type
  • Snaps closure type
Catalog No. 50-007-70254 Supplier MSC Supplier No. 06509186
This rain suit with pants is designed to provide non-hazardous protection. It features a yellow color and is constructed from durable polyester and PVC materials, suitable for various outdoor work environments.
  • Suit with pants garment style
  • Size: 3X-Large
  • Yellow color
  • Polyester and PVC material
  • Non-hazardous protection standards
  • Detachable hood
  • Bib pant type
  • Snaps closure type