Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Onion's Fusible Alloy
SCI_ED

Catalog No. S25776A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Metal
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25776A 100 g Metal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25776A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Specifications

Chemical Name or Material Onion's Fusible Alloy
CAS 7440-31-5, 76-61-9, 7439-92-1
Quantity 100 g
Packaging Metal
Assay Percent Range 20% w/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.