Learn More
Cayman Chemical OnjIsaponIn B 10mg
SDP
Catalog No. 502991348
Encompass_Preferred
A triterpenoid saponin with neuroprotective activity; inhibits glutamate- or serum-starvation-induced neurotoxicity in PC12 cells at 10 μM; decreases amyloid-β production in HEK293/APPswe cells (IC50 = 10 μM); reduces spatial learning and memory deficits in the Morris water maze in an APP/PS1 transgenic mouse model of Alzheimer’s disease at 10 mg/kg per day; prevents D-galactose-induced decreases in hippocampal GSH levels and increases in hippocampal MDA, IL-1β, IL-6, and TNF-α levels in a rat model of D-galactose-induced aging at 10 and 20 mg/kg
Catalog No. 50-299-1348 Supplier Cayman Chemical Supplier No. 3052110 MG
A triterpenoid saponin with neuroprotective activity; inhibits glutamate- or serum-starvation-induced neurotoxicity in PC12 cells at 10 μM; decreases amyloid-β production in HEK293/APPswe cells (IC50 = 10 μM); reduces spatial learning and memory deficits in the Morris water maze in an APP/PS1 transgenic mouse model of Alzheimer’s disease at 10 mg/kg per day; prevents D-galactose-induced decreases in hippocampal GSH levels and increases in hippocampal MDA, IL-1β, IL-6, and TNF-α levels in a rat model of D-galactose-induced aging at 10 and 20 mg/kg