Learn More
Medchemexpress LLC Onjisaponin B | 35906-36-6 | 99.1% | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN209910MG
Encompass_Preferred
Onjisaponin B is a natural triterpenoid saponin isolated from Polygala tenuifolia with reported neuroprotective and autophagy-enhancing activity. It is used in cell-based and preclinical studies investigating protein aggregation, neurodegeneration, and inflammatory signaling.
  • Natural triterpenoid saponin derived from Polygala tenuifolia.
  • Inhibits NF-κB p65 and modulates inflammatory signaling.
  • Enhances autophagy and promotes degradation of mutant α-synuclein and huntingtin.
  • High purity solid powder (99.1%).
  • Supplied as a 10 mg quantity suitable for research-scale assays.
  • Intended for mechanistic studies and preclinical pharmacology models.
Catalog No. 50-002-95943