Learn More
Medchemexpress LLC HY-N2099 5mg Medchemexpress, Onjisaponin B CAS:35906-36-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN20995MG
Encompass_Preferred
Medchemexpress, HY-N2099 5mg Onjisaponin B CAS:35906-36-6 Onjisaponin B is a natural product derived from Radix Polygalae. Onjisaponin B enhances autophagy and accelerates the degradation of mutant α-synuclein and huntingtin in PC-12 cells, and exbibits potential therapeutic effects on Parkinson disease and Huntington disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0366