Learn More
Hach Company Online Clean Water Dig ORP Sen
Catalog No. 5000207692 Shop All Hach Company Products
Encompass
Hach Online Process ORP Sensor - Clean Water Digital ORP SensorClean Water Online Process ORP Sensor with Integrated Digital Electronics for in Plug and Playin with Hach sc Digital Controllers - Platinum ORP Electrode PPS Housing Convertible Mount 10m Cable
Catalog No. 50-002-07692 Supplier Hach Company Supplier No. DRD1R5
Hach Online Process ORP Sensor - Clean Water Digital ORP SensorClean Water Online Process ORP Sensor with Integrated Digital Electronics for in Plug and Playin with Hach sc Digital Controllers - Platinum ORP Electrode PPS Housing Convertible Mount 10m Cable