Learn More
Hach Company Online GP Digital ORP Sensor
Catalog No. 5000207605 Shop All Hach Company Products
Encompass
Hach Online Process ORP Sensor - General Purpose Digital ORP SensorGeneral Purpose Online Process ORP Sensor with Integrated Digital Electronics for in Plug and Playin with Hach sc Digital Controllers - Platinum ORP Electrode PEEK Housing Convertible Mount 10m Cable
Catalog No. 50-002-07605 Supplier Hach Company Supplier No. DRD1P5
Hach Online Process ORP Sensor - General Purpose Digital ORP SensorGeneral Purpose Online Process ORP Sensor with Integrated Digital Electronics for in Plug and Playin with Hach sc Digital Controllers - Platinum ORP Electrode PEEK Housing Convertible Mount 10m Cable