Learn More
Hach Company Online GP SS ORP Sensor
Catalog No. 5000207683 Shop All Hach Company Products
Encompass
Hach Online Process ORP Sensor - General Purpose-Stainless Steel ORP SensorGeneral Purpose Online Process ORP Sensor with Integrated Digital Electronics for in Plug and Playin with Hach sc Digital Controllers - Platinum ORP Electrode Stainless Housing Immersion Mount 10m Cable
Catalog No. 50-002-07683 Supplier Hach Company Supplier No. DRS5
Hach Online Process ORP Sensor - General Purpose-Stainless Steel ORP SensorGeneral Purpose Online Process ORP Sensor with Integrated Digital Electronics for in Plug and Playin with Hach sc Digital Controllers - Platinum ORP Electrode Stainless Housing Immersion Mount 10m Cable