Learn More
Perkin Elmer US LLC Online Mixing Kit for MEINHARD Plus Nebulizers, 0.5 mm I.D. - Orange
Catalog No. 502104229
Encompass_Preferred
Includes: Low dead-volume, fluoropolymer mixing tee 0.5 mm I.D. (Orange) for flow rates > 1 mL/min Internal standard probe and addition line Fluoropolymer peripump fittings Peristaltic pump tubing starter pack for the MP2 micro peristaltic pump Note: The PFA Adapter for Glass nebulizers (N8145365) allows this kit to be used with other types of glass nebulizers.
Catalog No. 50-210-4229 Supplier Perkin Elmer US LLC Supplier No. N8145361
Includes: Low dead-volume, fluoropolymer mixing tee 0.5 mm I.D. (Orange) for flow rates > 1 mL/min Internal standard probe and addition line Fluoropolymer peripump fittings Peristaltic pump tubing starter pack for the MP2 micro peristaltic pump Note: The PFA Adapter for Glass nebulizers (N8145365) allows this kit to be used with other types of glass nebulizers.