Learn More
Hach Company Online pH Sensor HF Res. Low
Catalog No. 5000207283 Shop All Hach Company Products
Encompass
Hach pHD Online Process pH Sensor - pH Sensor for HF Resistance-Low pHHF Resistant Low pH Online Process pH Sensor - pHD Technology Glass pH Electrode PEEK Housing Insertion Mount 4 5m Cable
Catalog No. 50-002-07283 Supplier Hach Company Supplier No. PD2P3
Hach pHD Online Process pH Sensor - pH Sensor for HF Resistance-Low pHHF Resistant Low pH Online Process pH Sensor - pHD Technology Glass pH Electrode PEEK Housing Insertion Mount 4 5m Cable