Learn More
Hach Company Online pH Sensor Incert Mt.
Catalog No. 5000208088 Shop All Hach Company Products
Encompass
Hach pHD sc Online Process pH Sensor - General Purpose Insertion Mount pH SensorGeneral Purpose Online Process pH Sensor with Integrated Digital Electronics for in Plug and Playin with Hach Digital SC Controllers - pHD Technology Glass pH Electrode PEEK Housing Insertion Mount 10 m CableThis instrument connects to Claros Hach s innovative Water Intelligence System enabling you to seamlessly connect and manage instruments data and process anywhere anytime The result is greater confidence in your data and improved efficiency in your operations To unlock the full potential of Claros insist on Claros Enabled instruments
This item is not returnable. View return policy
Catalog No. 50-002-08088 Supplier Hach Company Supplier No. DPD2P1
Hach pHD sc Online Process pH Sensor - General Purpose Insertion Mount pH SensorGeneral Purpose Online Process pH Sensor with Integrated Digital Electronics for in Plug and Playin with Hach Digital SC Controllers - pHD Technology Glass pH Electrode PEEK Housing Insertion Mount 10 m CableThis instrument connects to Claros Hach s innovative Water Intelligence System enabling you to seamlessly connect and manage instruments data and process anywhere anytime The result is greater confidence in your data and improved efficiency in your operations To unlock the full potential of Claros insist on Claros Enabled instruments