Learn More
Hach Company Online pH Sensor San. Mt.
Catalog No. 5000207784 Shop All Hach Company Products
Encompass
Hach pHD sc Online Process pH Sensor - General Purpose Sanitary Mount pH SensorGeneral Purpose Online Process pH Sensor with Integrated Digital Electronics for in Plug and Playin with Hach Digital SC Controllers - pHD Technology Glass pH Electrode PEEK Housing Sanitary Mount 10 m CableThis instrument connects to Claros Hach s innovative Water Intelligence System enabling you to seamlessly connect and manage instruments data and process anywhere anytime The result is greater confidence in your data and improved efficiency in your operations To unlock the full potential of Claros insist on Claros Enabled instruments
Catalog No. 50-002-07784 Supplier Hach Company Supplier No. DPD3P1
Hach pHD sc Online Process pH Sensor - General Purpose Sanitary Mount pH SensorGeneral Purpose Online Process pH Sensor with Integrated Digital Electronics for in Plug and Playin with Hach Digital SC Controllers - pHD Technology Glass pH Electrode PEEK Housing Sanitary Mount 10 m CableThis instrument connects to Claros Hach s innovative Water Intelligence System enabling you to seamlessly connect and manage instruments data and process anywhere anytime The result is greater confidence in your data and improved efficiency in your operations To unlock the full potential of Claros insist on Claros Enabled instruments