Learn More
Hach Company Online Process pH Sensor SS
Catalog No. 5000207789 Shop All Hach Company Products
Encompass
Hach pHD sc Online Process pH Sensor - General Purpose Stainless Steel pH SensorGeneral Purpose Online Process pH Sensor with Integrated Digital Electronics for in Plug and Playin with Hach Digital SC Controllers - pHD Technology Glass pH Electrode Stainless Housing Immersion Mount 10 m CableThis instrument connects to Claros Hach s innovative Water Intelligence System enabling you to seamlessly connect and manage instruments data and process anywhere anytime The result is greater confidence in your data and improved efficiency in your operations To unlock the full potential of Claros insist on Claros Enabled instruments
Catalog No. 50-002-07789 Supplier Hach Company Supplier No. DPS1
Hach pHD sc Online Process pH Sensor - General Purpose Stainless Steel pH SensorGeneral Purpose Online Process pH Sensor with Integrated Digital Electronics for in Plug and Playin with Hach Digital SC Controllers - pHD Technology Glass pH Electrode Stainless Housing Immersion Mount 10 m CableThis instrument connects to Claros Hach s innovative Water Intelligence System enabling you to seamlessly connect and manage instruments data and process anywhere anytime The result is greater confidence in your data and improved efficiency in your operations To unlock the full potential of Claros insist on Claros Enabled instruments