Learn More
Hach Company Online Special Dig. ORP Sensor
Catalog No. 5000207788 Shop All Hach Company Products
Encompass
Hach Online Process ORP Sensor - Speciality Digital ORP SensorSpecialty Online Process ORP Sensor with Integrated Digital Electronics for in Plug and Playin with Hach sc Digital Controllers - Gold ORP Electrode PEEK Housing Convertible Mount 10m Cable
Catalog No. 50-002-07788 Supplier Hach Company Supplier No. DRD1P6
Hach Online Process ORP Sensor - Speciality Digital ORP SensorSpecialty Online Process ORP Sensor with Integrated Digital Electronics for in Plug and Playin with Hach sc Digital Controllers - Gold ORP Electrode PEEK Housing Convertible Mount 10m Cable