Learn More
Medchemexpress LLC ONO-0300302 10mg | 856689-51-5 | 10 MG
SDP
Catalog No. 5000229750
Encompass_Preferred
ONO-0300302 is an orally active, potent lysophosphatidic acid receptor 1 (LPA1) antagonist used in preclinical research, including models of benign prostatic hyperplasia. It is a slow tight-binding inhibitor with reported IC50 0.086 μM and Kd 0.34 nM, and is supplied at high purity for experimental use.
  • Orally active LPA1 antagonist with IC50 0.086 μM.
  • Slow, tight-binding inhibitor with a reported Kd of 0.34 nM (37 °C, 2 h).
  • High supplied purity, suitable for research applications.
  • Available in small pack sizes for preclinical experiments.
  • Chemical formula C29H35NO5; molecular weight 477.59 g·mol⁻1.
Catalog No. 50-002-29750 Supplier Medchemexpress LLC Supplier No. HY11545010MG
ONO-0300302 is an orally active, potent lysophosphatidic acid receptor 1 (LPA1) antagonist used in preclinical research, including models of benign prostatic hyperplasia. It is a slow tight-binding inhibitor with reported IC50 0.086 μM and Kd 0.34 nM, and is supplied at high purity for experimental use.
  • Orally active LPA1 antagonist with IC50 0.086 μM.
  • Slow, tight-binding inhibitor with a reported Kd of 0.34 nM (37 °C, 2 h).
  • High supplied purity, suitable for research applications.
  • Available in small pack sizes for preclinical experiments.
  • Chemical formula C29H35NO5; molecular weight 477.59 g·mol⁻1.