Learn More
Medchemexpress LLC ONO-0300302 50mg | 856689-51-5 | 477.59 | C29H35NO5 | 50 MG
SDP
Catalog No. 5000226460
Encompass_Preferred
ONO-0300302 is an orally active, potent lysophosphatidic acid receptor 1 (LPA1) antagonist used in pharmacological research. It functions as a slow, tight-binding inhibitor (IC50 = 0.086 μM; Kd = 0.34 nM at 37 °C, 2 h) and is supplied with high reported purity for in vitro and preclinical studies.
  • Potent LPA1 antagonist (IC50 = 0.086 μM).
  • Slow, tight-binding kinetics with time-dependent increase in affinity.
  • Reported purity 99.43%.
  • Molecular formula C29H35NO5; molecular weight 477.59.
  • Available as solids (5 mg-100 mg; larger sizes by quote) and as 10 mM solution in DMSO.
Catalog No. 50-002-26460 Supplier Medchemexpress LLC Supplier No. HY11545050MG
ONO-0300302 is an orally active, potent lysophosphatidic acid receptor 1 (LPA1) antagonist used in pharmacological research. It functions as a slow, tight-binding inhibitor (IC50 = 0.086 μM; Kd = 0.34 nM at 37 °C, 2 h) and is supplied with high reported purity for in vitro and preclinical studies.
  • Potent LPA1 antagonist (IC50 = 0.086 μM).
  • Slow, tight-binding kinetics with time-dependent increase in affinity.
  • Reported purity 99.43%.
  • Molecular formula C29H35NO5; molecular weight 477.59.
  • Available as solids (5 mg-100 mg; larger sizes by quote) and as 10 mM solution in DMSO.