Learn More
Medchemexpress LLC ONO-2952 | 895169-20-7 | 99.5% | 398.86 | 10 MG
SDP
Catalog No. 5000343416
Encompass_Preferred
ONO-2952 is a potent, selective, and orally active translocator protein 18 kDa (TSPO) antagonist. It is more selective for TSPO than other receptors, transporters, ion channels, and enzymes. It exerts anti-stress effects by inhibiting excessive activation of the noradrenergic system in the brain without causing amnesic effects. This product has potential for irritable bowel syndrome treatment and is for research use only.
  • Potent, selective, and orally active translocator protein 18 kDa (TSPO) antagonist with Ki of 0.33-9.30 nM for rat and human TSPO.
  • More selective for TSPO than other receptors, transporters, ion channels, and enzymes.
  • Exerts anti-stress effects through inhibition of excessive activation of the noradrenergic system in the brain without the amnesic effect.
  • Potential for irritable bowel syndrome treatment.
Catalog No. 50-003-43416 Supplier Medchemexpress LLC Supplier No. HY11119110MG
ONO-2952 is a potent, selective, and orally active translocator protein 18 kDa (TSPO) antagonist. It is more selective for TSPO than other receptors, transporters, ion channels, and enzymes. It exerts anti-stress effects by inhibiting excessive activation of the noradrenergic system in the brain without causing amnesic effects. This product has potential for irritable bowel syndrome treatment and is for research use only.
  • Potent, selective, and orally active translocator protein 18 kDa (TSPO) antagonist with Ki of 0.33-9.30 nM for rat and human TSPO.
  • More selective for TSPO than other receptors, transporters, ion channels, and enzymes.
  • Exerts anti-stress effects through inhibition of excessive activation of the noradrenergic system in the brain without the amnesic effect.
  • Potential for irritable bowel syndrome treatment.