Learn More
Medchemexpress LLC Ono-2952 | 895169-20-7 | MFCD32661907 | 99.5% | 398.9 g/mol | C22H20ClFN2O2 | 5 MG
SDP
Catalog No. 5000263585
Encompass_Preferred
ONO-2952 is a potent, selective, orally active translocator protein 18 kDa (TSPO) antagonist used in pharmacology research. It displays low-nanomolar binding affinity to rat and human TSPO and has been evaluated in studies related to gastrointestinal pain and irritable bowel syndrome.
  • Potent TSPO antagonist with Ki in the low nanomolar range.
  • High chemical purity (≈99.5%).
  • Molecular formula C22H20ClFN2O2; mol wt 398.9 g/mol.
  • Available as solid and as 10 mM solution in DMSO for stock preparation.
  • Supplied in small research quantities suitable for assay development.
  • Characterized for identity (CAS 895169-20-7) with supporting spectral data.
Catalog No. 50-002-63585 Supplier Medchemexpress LLC Supplier No. HY1111915MG
ONO-2952 is a potent, selective, orally active translocator protein 18 kDa (TSPO) antagonist used in pharmacology research. It displays low-nanomolar binding affinity to rat and human TSPO and has been evaluated in studies related to gastrointestinal pain and irritable bowel syndrome.
  • Potent TSPO antagonist with Ki in the low nanomolar range.
  • High chemical purity (≈99.5%).
  • Molecular formula C22H20ClFN2O2; mol wt 398.9 g/mol.
  • Available as solid and as 10 mM solution in DMSO for stock preparation.
  • Supplied in small research quantities suitable for assay development.
  • Characterized for identity (CAS 895169-20-7) with supporting spectral data.