Learn More
Medchemexpress LLC Ono-5334 | 868273-90-9 | 99.8% | 438.58 g/mol | C21H34N4O4S | 1 MG
SDP
Catalog No. 5000242985
Encompass_Preferred
ONO-5334 is a small-molecule, orally active cathepsin K inhibitor used as a research compound to study bone resorption, osteoclast activity, and related biochemical pathways. It has been evaluated in preclinical and clinical settings and is commonly employed in biochemical assays and cell-based studies to probe cathepsin-mediated mechanisms.
  • Potent, selective cathepsin K inhibition for target-focused studies.
  • Orally active small molecule suitable for in vitro and in vivo models.
  • Useful for investigating bone resorption and osteoclast function.
  • Available in low-mass vials for research dosing and assay work.
  • High purity compatible with analytical and biological experiments.
Catalog No. 50-002-42985 Supplier Medchemexpress LLC Supplier No. HY1080441MG
ONO-5334 is a small-molecule, orally active cathepsin K inhibitor used as a research compound to study bone resorption, osteoclast activity, and related biochemical pathways. It has been evaluated in preclinical and clinical settings and is commonly employed in biochemical assays and cell-based studies to probe cathepsin-mediated mechanisms.
  • Potent, selective cathepsin K inhibition for target-focused studies.
  • Orally active small molecule suitable for in vitro and in vivo models.
  • Useful for investigating bone resorption and osteoclast function.
  • Available in low-mass vials for research dosing and assay work.
  • High purity compatible with analytical and biological experiments.