Learn More
Medchemexpress LLC Ono-8430506 | 1354805-08-5 | 99.9% | C27H28FN3O3 | 50 MG
SDP
Catalog No. 5000354343
Encompass_Preferred
This orally bioavailable and potent autotaxin (ATX)/ENPP2 inhibitor efficiently inhibits lysophosphatidic acid (LPA) formation with IC50s of approximately 10 nM. In vivo, it slows initial tumor growth, limits lung metastasis in mice, and enhances the antitumor effect of Paclitaxel in a breast cancer model. This product is for research use only.
  • Potent autotaxin (ATX)/ENPP2 inhibitor
  • Orally bioavailable
  • Efficiently inhibits lysophosphatidic acid (LPA) formation
  • Slows initial tumor growth and limits lung metastasis
  • Enhances antitumor effect of Paclitaxel in breast cancer models
Catalog No. 50-003-54343 Supplier Medchemexpress LLC Supplier No. HY14534450MG
This orally bioavailable and potent autotaxin (ATX)/ENPP2 inhibitor efficiently inhibits lysophosphatidic acid (LPA) formation with IC50s of approximately 10 nM. In vivo, it slows initial tumor growth, limits lung metastasis in mice, and enhances the antitumor effect of Paclitaxel in a breast cancer model. This product is for research use only.
  • Potent autotaxin (ATX)/ENPP2 inhibitor
  • Orally bioavailable
  • Efficiently inhibits lysophosphatidic acid (LPA) formation
  • Slows initial tumor growth and limits lung metastasis
  • Enhances antitumor effect of Paclitaxel in breast cancer models