Learn More
Medchemexpress LLC Ono-9780307 | 856691-44-6 | 99.9% | 521.6 g/mol | C30H35NO7 | 10 MG
SDP
Catalog No. 5000226999
Encompass_Preferred
ONO-9780307 is a synthetic antagonist of lysophosphatidic acid receptor 1 (LPA1) used as a research tool. It exhibits reported potency (IC50 ≈ 2.7 nM), has molecular formula C30H35NO7, and molecular weight 521.6 g/mol. Supplied as a high-purity research sample for in vitro and structural biology studies; not for human or veterinary use.
  • Specific LPA1 antagonist with reported IC50 ≈ 2.7 nM.
  • Used in structural biology and pharmacology research.
  • High purity suitable for biochemical and biophysical assays.
  • Provided as a 10 mg solid for laboratory use.
  • For research use only; not for human or veterinary administration.
Catalog No. 50-002-26999 Supplier Medchemexpress LLC Supplier No. HY11744410MG
ONO-9780307 is a synthetic antagonist of lysophosphatidic acid receptor 1 (LPA1) used as a research tool. It exhibits reported potency (IC50 ≈ 2.7 nM), has molecular formula C30H35NO7, and molecular weight 521.6 g/mol. Supplied as a high-purity research sample for in vitro and structural biology studies; not for human or veterinary use.
  • Specific LPA1 antagonist with reported IC50 ≈ 2.7 nM.
  • Used in structural biology and pharmacology research.
  • High purity suitable for biochemical and biophysical assays.
  • Provided as a 10 mg solid for laboratory use.
  • For research use only; not for human or veterinary administration.