Learn More
Medchemexpress LLC Ono-9780307 | 856691-44-6 | 99.9% | 521.60 g·mol-1 | C30H35NO7 | 5 MG
SDP
Catalog No. 5000226737
Encompass_Preferred
ONO-9780307 is a synthetic LPA1 (lysophosphatidic acid receptor 1) antagonist supplied for research use and structural studies. It is commonly used in receptor pharmacology assays and to support crystallography and other biophysical investigations.
  • Antagonist of LPA1 with reported IC50 ≈ 2.7 nM.
  • High purity suitable for biochemical assays (vendor-reported ~99.9%).
  • Molecular formula C30H35NO7 and molecular weight 521.60 g·mol-1.
  • Supplied as a solid, typical pack size 5 mg for laboratory use.
  • Intended for research use only; not for human or veterinary applications.
Catalog No. 50-002-26737 Supplier Medchemexpress LLC Supplier No. HY1174445MG
ONO-9780307 is a synthetic LPA1 (lysophosphatidic acid receptor 1) antagonist supplied for research use and structural studies. It is commonly used in receptor pharmacology assays and to support crystallography and other biophysical investigations.
  • Antagonist of LPA1 with reported IC50 ≈ 2.7 nM.
  • High purity suitable for biochemical assays (vendor-reported ~99.9%).
  • Molecular formula C30H35NO7 and molecular weight 521.60 g·mol-1.
  • Supplied as a solid, typical pack size 5 mg for laboratory use.
  • Intended for research use only; not for human or veterinary applications.