Learn More
Enzo Life Sciences ONO-RS-082 (20 mg). CAS: 99754-06-0
SDP
Supplier:  Enzo Life Sciences BMLST3200020
Encompass_Preferred
Phospholipase A2 inhibitor (IC50=7 µM for purified guinea pig lung PLA2). Inhibits epinephrine-stimulated thromboxane production in human platelets (86% at 3.5µM). Also blocks insulin secretion in pancreatic islets in response to glucose. Disrupts membrane tubule-mediated reassembly and maintenance of the Golgi complex. Alternative name: 2-(p-Amylcinnamoyl)amino-4-chlorobenzoic acid. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml, warm) or 100% ethanol (10mg/ml, warm). Long Term Storage: Ambient.
Catalog No. 50-201-2156