Learn More
Medchemexpress LLC Ononetin | 487-49-0 | 99.5% | 258.27 | 50 MG
SDP
Catalog No. 5000346973
Encompass_Preferred
Ononetin, a natural deoxybenzoin, is a potent and selective TRPM3 channel blocker with an IC50 of 0.3 μM.
  • Completely and reversibly abrogates Ca2+ entry and ionic currents through recombinantly expressed TRPM3α2.
  • Blocks pregnenolone sulphate-inducible Ca2+ entry in primary cultures of mouse or rat dorsal root ganglia (DRG) neurones.
  • Reverses established Freund's Complete Adjuvant (FCA)-induced heat hypersensitivity.
  • Suggests TRPM3 may be a tractable target for inflammatory pain.
Catalog No. 50-003-46973 Supplier Medchemexpress LLC Supplier No. HY10845110MG
Ononetin, a natural deoxybenzoin, is a potent and selective TRPM3 channel blocker with an IC50 of 0.3 μM.
  • Completely and reversibly abrogates Ca2+ entry and ionic currents through recombinantly expressed TRPM3α2.
  • Blocks pregnenolone sulphate-inducible Ca2+ entry in primary cultures of mouse or rat dorsal root ganglia (DRG) neurones.
  • Reverses established Freund's Complete Adjuvant (FCA)-induced heat hypersensitivity.
  • Suggests TRPM3 may be a tractable target for inflammatory pain.