Learn More
Medchemexpress LLC HY-15926 5g , ONPG 2-Nitrophenyl β-D-galactopyranoside CAS:369-07-3 Purity:98%
SDP
Catalog No. 501492210
Encompass_Preferred
Medchemexpress, HY-15926 5g ONPG 2-Nitrophenyl β-D-galactopyranoside CAS:369-07-3 Formula:C12H15NO8 Purity:98% ONPG is a colorimetric and spectrophotometric substrate for detection of β-galactosidase activity. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2210 Supplier Medchemexpress LLC Supplier No. HY159265G
Medchemexpress, HY-15926 5g ONPG 2-Nitrophenyl β-D-galactopyranoside CAS:369-07-3 Formula:C12H15NO8 Purity:98% ONPG is a colorimetric and spectrophotometric substrate for detection of β-galactosidase activity. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.