Learn More
Alkali Scientific [ONPG] 2 Nitrophenyl β D Galactopyranoside, 1 Gram
SDP
Catalog No. 5000661049
Encompass_Preferred
2-Nitrophenyl β-D-galactopyranoside is a reagent supplied in a 1g quantity, used in enzymatic assays to study the activity of β-galactosidase. This compound serves as a substrate for the hydrolysis of the β-galactosidic bond, a key reaction catalyzed by β-galactosidase. The reagent is commonly used in research on enzyme kinetics, carbohydrate metabolism, and the role of β-galactosidase in the breakdown of lactose and other galactose-containing compounds. This substrate is particularly useful in diagnostic assays and biochemical studies focusing on enzyme activity and substrate specificity, providing valuable insights into glycosidic bond cleavage.
Catalog No. 50-006-61049 Supplier Alkali Scientific Supplier No. ONP003CP
2-Nitrophenyl β-D-galactopyranoside is a reagent supplied in a 1g quantity, used in enzymatic assays to study the activity of β-galactosidase. This compound serves as a substrate for the hydrolysis of the β-galactosidic bond, a key reaction catalyzed by β-galactosidase. The reagent is commonly used in research on enzyme kinetics, carbohydrate metabolism, and the role of β-galactosidase in the breakdown of lactose and other galactose-containing compounds. This substrate is particularly useful in diagnostic assays and biochemical studies focusing on enzyme activity and substrate specificity, providing valuable insights into glycosidic bond cleavage.