Learn More
Abcam oNPG, beta-galactosidase substrate, 1G
Catalog No. 502704661
Encompass_Preferred
MW 301.25 g/mol. β-galactosidase substrate for colorimetric and EIA applications. Hydrolysed by β-galactosidase to yield galactose and ortho-Nitrophenol, which is yellow in color.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4661 Supplier Abcam Supplier No. AB1463921G
MW 301.25 g/mol. β-galactosidase substrate for colorimetric and EIA applications. Hydrolysed by β-galactosidase to yield galactose and ortho-Nitrophenol, which is yellow in color.
The product is subject to the following: Abcam Restricted Use Statement