Learn More
Andwin Scientific ONSET HOBO PRO V2
SDP
Catalog No. NC1073926
Encompass
The HOBO Water Temperature Pro v2 Data Logger is able to measure temperature within the range of -40C and 70C (-40F to 158F) in air and up to 50C (122F) in water. It can also work in either fresh or salt water. This product features an optical USB interface that makes it possible to offload and read data even while the logger is wet or underwater. It is waterproof to 120 meters (400 feet). This can work with HOBOware and HOBOware Pro software for logger setup, graphing and analysis.
Catalog No. NC1073926 Supplier Andwin Scientific Supplier No. U22001
The HOBO Water Temperature Pro v2 Data Logger is able to measure temperature within the range of -40C and 70C (-40F to 158F) in air and up to 50C (122F) in water. It can also work in either fresh or salt water. This product features an optical USB interface that makes it possible to offload and read data even while the logger is wet or underwater. It is waterproof to 120 meters (400 feet). This can work with HOBOware and HOBOware Pro software for logger setup, graphing and analysis.