Learn More
Medchemexpress LLC Ontunisertib (AGMB-129) | 2647949-48-0 | 99.95% | 469.49 | 5 MG
SDP
Catalog No. 5000390869
Encompass_Preferred
Ontunisertib (AGMB-129) is an orally active and selective gastrointestinal-restricted ALK5 (TGFβR1) inhibitor. It blocks signaling of the pro-fibrotic TGFβ pathway and can be used for the research of fibrostenotic Crohn's disease.
  • Orally active and selective gastrointestinal-restricted ALK5 (TGFβR1) inhibitor.
  • Blocks signaling of the pro-fibrotic TGFβ pathway.
  • Can be used for the research of fibrostenotic Crohn's disease.
  • Designed to act locally in the gastrointestinal tract, enabling high exposure in the target tissue.
  • Rapidly inactivated in the liver following absorption to avoid potential toxicities associated with systemic TGFβ signaling inhibition.
Catalog No. 50-003-90869 Supplier Medchemexpress LLC Supplier No. HY1689905MG
Ontunisertib (AGMB-129) is an orally active and selective gastrointestinal-restricted ALK5 (TGFβR1) inhibitor. It blocks signaling of the pro-fibrotic TGFβ pathway and can be used for the research of fibrostenotic Crohn's disease.
  • Orally active and selective gastrointestinal-restricted ALK5 (TGFβR1) inhibitor.
  • Blocks signaling of the pro-fibrotic TGFβ pathway.
  • Can be used for the research of fibrostenotic Crohn's disease.
  • Designed to act locally in the gastrointestinal tract, enabling high exposure in the target tissue.
  • Rapidly inactivated in the liver following absorption to avoid potential toxicities associated with systemic TGFβ signaling inhibition.