Learn More
Selleck Chemical LLC ONX-0914
SDP
Catalog No. 5000000827
Encompass_Preferred
ONX-0914 (PR-957) is a potent and selective immunoproteasome inhibitor with minimal cross-reactivity for the constitutive proteasome in a cell-free assay
Catalog No. 50-000-00827 Supplier Selleck Chemical LLC Supplier No. S717225MG
ONX-0914 (PR-957) is a potent and selective immunoproteasome inhibitor with minimal cross-reactivity for the constitutive proteasome in a cell-free assay