Learn More
Medchemexpress LLC Onx-0914 trifluoroacetate | 00-00-0 | MFCD26794217 | 98.0% | 694.70 | C33H41F3N4O9 | 5 MG
SDP
Catalog No. 5000262369
Encompass_Preferred
ONX-0914 trifluoroacetate is the TFA salt of a selective immunoproteasome LMP7 (β5i) inhibitor used in research to modulate proteasome activity and investigate inflammation and autoimmune pathways. It is supplied as a solid and as DMSO solutions for flexible experimental use; molecular weight and formula correspond to the TFA salt form.
  • Selective inhibition of the LMP7 immunoproteasome subunit
  • Irreversible, noncompetitive mechanism for pathway interrogation
  • Available as solid and DMSO solution for flexible dosing
  • High reported purity suitable for biochemical assays
  • Soluble in DMSO and compatible with cell-based studies
Catalog No. 50-002-62369 Supplier Medchemexpress LLC Supplier No. HY13207A5MG
ONX-0914 trifluoroacetate is the TFA salt of a selective immunoproteasome LMP7 (β5i) inhibitor used in research to modulate proteasome activity and investigate inflammation and autoimmune pathways. It is supplied as a solid and as DMSO solutions for flexible experimental use; molecular weight and formula correspond to the TFA salt form.
  • Selective inhibition of the LMP7 immunoproteasome subunit
  • Irreversible, noncompetitive mechanism for pathway interrogation
  • Available as solid and DMSO solution for flexible dosing
  • High reported purity suitable for biochemical assays
  • Soluble in DMSO and compatible with cell-based studies