Learn More
Smooth On Inc OOMOO™ 30 Silcon Rubber, Trial Size Unit (2.8 lbs. / 1.27 kg.)
SDP
Catalog No. NC0610334
Encompass
OOMOO™ 30No Vacuuming – No Scale – Easy To Use . . . OOMOO™ 30 is an easy to use tin cure silicone rubber compound that features a convenient one-to-one by volume mix ratio (no scale necessary). It has low viscosity for easy mixing and pouring . . . vacuum degassing is not necessary. This product cures at room temperature with negligible shrinkage. OOMOO™ 30 has a 30-minute pot life, with a six-hour cure time.
Catalog No. NC0610334 Supplier Smooth On Inc Supplier No. 82141
OOMOO™ 30No Vacuuming – No Scale – Easy To Use . . . OOMOO™ 30 is an easy to use tin cure silicone rubber compound that features a convenient one-to-one by volume mix ratio (no scale necessary). It has low viscosity for easy mixing and pouring . . . vacuum degassing is not necessary. This product cures at room temperature with negligible shrinkage. OOMOO™ 30 has a 30-minute pot life, with a six-hour cure time.