Learn More
Medchemexpress LLC OP-5244 sodium | 00-00-0 | 99.9% | 559.87 g/mol | C19H28ClN5NaO9P | 5 MG
SDP
Catalog No. 5000222348
Encompass_Preferred
OP-5244 sodium is the sodium salt form of a potent, orally active CD73 (ecto-5′-nucleotidase) inhibitor used as a biochemical probe in cancer and immunology research. It blocks adenosine production to reverse immunosuppression and is supplied as a high-purity solid suitable for in vitro and in vivo studies.
  • Potent CD73 inhibition (IC50 0.25 nM).
  • Sodium salt formulation for improved aqueous solubility.
  • High purity (99.87%).
  • Small research-scale sizes available (e.g., 5 mg).
  • Store sealed at 4°C; follow recommended handling for solutions.
Catalog No. 50-002-22348 Supplier Medchemexpress LLC Supplier No. HY136978A5MG
OP-5244 sodium is the sodium salt form of a potent, orally active CD73 (ecto-5′-nucleotidase) inhibitor used as a biochemical probe in cancer and immunology research. It blocks adenosine production to reverse immunosuppression and is supplied as a high-purity solid suitable for in vitro and in vivo studies.
  • Potent CD73 inhibition (IC50 0.25 nM).
  • Sodium salt formulation for improved aqueous solubility.
  • High purity (99.87%).
  • Small research-scale sizes available (e.g., 5 mg).
  • Store sealed at 4°C; follow recommended handling for solutions.