Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Op-5244 sodium | 00-00-0 | 99.9% | C19H28ClN5NaO9P | 10MG
SDP

Catalog No. 5000261260
Encompass_Preferred

OP-5244 sodium is the sodium salt of a potent, orally active small-molecule CD73 inhibitor that reverses immunosuppression by blocking adenosine production. Supplied for research use as a characterized solid with accompanying purity and analytical data.

  • Potent CD73 inhibitor (IC50 0.25 nM).
  • Reverses adenosine-mediated immunosuppression.
  • High purity (99.9%).
  • Chemical formula C19H28ClN5NaO9P; molecular weight 559.87 g·mol⁻¹.
  • Available in small research quantities such as 10 mg.
  • Solid form; store sealed at 4 °C.

Catalog No. 50-002-61260 Supplier Medchemexpress LLC Supplier No. HY136978A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

OP-5244 sodium is the sodium salt of a potent, orally active small-molecule CD73 inhibitor that reverses immunosuppression by blocking adenosine production. Supplied for research use as a characterized solid with accompanying purity and analytical data.

  • Potent CD73 inhibitor (IC50 0.25 nM).
  • Reverses adenosine-mediated immunosuppression.
  • High purity (99.9%).
  • Chemical formula C19H28ClN5NaO9P; molecular weight 559.87 g·mol⁻¹.
  • Available in small research quantities such as 10 mg.
  • Solid form; store sealed at 4 °C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.