Learn More
Roboz Surgical OP SCISSORS STR CRBIDE 5 1/2IN
SDP
Catalog No. NC9585525
Encompass
These 5 1/2-inch straight operating scissors are precision-engineered for reliable cutting in surgical and laboratory applications. Designed with a sharp-blunt tip configuration, they provide versatility for both delicate dissection and general utility tasks.
  • Tungsten carbide inserts on the blades provide enhanced edge retention and cutting performance.
  • Straight design allows for precise access in controlled environments.
  • Sharp-blunt tip configuration offers utility for diverse surgical procedures.
  • Constructed for durability and consistent performance in demanding clinical settings.
Catalog No. NC9585525 Supplier Roboz Surgical Supplier No. RS6832
These 5 1/2-inch straight operating scissors are precision-engineered for reliable cutting in surgical and laboratory applications. Designed with a sharp-blunt tip configuration, they provide versatility for both delicate dissection and general utility tasks.
  • Tungsten carbide inserts on the blades provide enhanced edge retention and cutting performance.
  • Straight design allows for precise access in controlled environments.
  • Sharp-blunt tip configuration offers utility for diverse surgical procedures.
  • Constructed for durability and consistent performance in demanding clinical settings.